Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Harvard Apparatus™ Large Nylon Clamps

$21.00

Specifications

Size Large
For Use With (Equipment) BTX Harvard Apparatus
Material Nylon
Type Stronghold Clamp
Length (English) Opening 3.33 in.
View More Specs

Products 2
Catalog Number Mfr. No. Color Includes Price Quantity & Availability  
Catalog Number Mfr. No. Color Includes Price Quantity & Availability  
05-886-11
View Documents
Harvard Apparatus™
53 2112R
Red Extension stem
Each for $21.00
Add to cart
 
05-886-10
View Documents
Harvard Apparatus™
53 2110W
White Male connector This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

Two moving jaws open to 31/3 in. (85mm). Close to within 1/8 in. (3mm) of contact. Made of strong, lightweight nylon reinforced with glass fiber. Resist most chemicals, particularly salt solutions and solvents. Heat resistant to 221°C (430°F). Choice of male connector or extension stem. Extension stem is 3/8 in. diameter (9.5mm) and 6 in. long (150mm). Choose red or white; integral color won't peel or flake.

Specifications

Specifications

Large
Nylon
3.33 in.
430°F
BTX Harvard Apparatus
Stronghold Clamp
8.45 cm
221°C
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit