Learn More
Brady™ Large Self-Laminating Blank Tags
Prevent your text from smearing or smudging in some of the harshest environments. Brady™ Large Self-Laminating Blank Tags are durable one-sided tags with a write-on area that accepts most marking pens.
Manufacturer:  Brady™ 102010
 View more versions of this product
Catalog No. 19-112-951
Add to cart
Description
Description
  • Polyester tags provide identification solutions for almost any environment
  • Flexible and versatile tags perform multiple roles in URD and related cable marking
  • In range of colors, stock sizes and hold configurations, these 10 mil. polyester tags flex and conform easily to cables of any size
  • Tag includes a write-on area that accepts most marking pens
  • Tags are appropriate for indoor or outdoor use
  • Clear, self-laminating flap protects written information from water
Specifications
Specifications
Yellow
(25) nylon fasteners
Rectangle
Polyester
3.25 in. x 5 in.
25/Pack
Water-resistant