Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LARP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5117002
Description
Positive Samples: HeLa, MCF7 Immunogen sequence: AGAAGAGRRD FVEAPPPKVN PWTKNALPPV LTTVNGQSPP EHSAPAKVVR AAVPKQRKGS KVGDFGDAIN WPTPGEIAHK SVQPQSHKPQ PTRKLPPKKD.
RNA-binding protein that regulates the translation of specific target mRNA species downstream of the mTORC1 complex, in function of growth signals and nutrient availability (PubMed:20430826, PubMed:23711370, PubMed:24532714, PubMed:25940091, PubMed:28650797, PubMed:28673543, PubMed:29244122). Interacts on the one hand with the 3' poly-A tails that are present in all mRNA molecules, and on the other hand with the 7-methylguanosine cap structure of mRNAs containing a 5' terminal oligopyrimidine (5'TOP) motif, which is present in mRNAs encoding ribosomal proteins and several components of the translation machinery (PubMed:23711370, PubMed:25940091, PubMed:28650797, PubMed:29244122, PubMed:26206669, PubMed:28379136). The interaction with the 5' end of mRNAs containing a 5'TOP motif leads to translational repression by preventing the binding of EIF4G1 (PubMed:25940091, PubMed:28650797, PubMed:29244122, PubMed:28379136). When mTORC1 is activated, LARP1 is phosphorylated and dissociates from the 5' untranslated region (UTR) of mRNA (PubMed:25940091, PubMed:28650797). Does not prevent binding of EIF4G1 to mRNAs that lack a 5'TOP motif (PubMed:28379136). Interacts with the free 40S ribosome subunit and with ribosomes, both monosomes and polysomes (PubMed:20430826, PubMed:24532714, PubMed:25940091, PubMed:28673543). Under normal nutrient availability, interacts primarily with the 3' untranslated region (UTR) of mRNAs encoding ribosomal proteins and increases protein synthesis (PubMed:23711370, PubMed:28650797). Associates with actively translating ribosomes and stimulates translation of mRNAs containing a 5'TOP motif, thereby regulating protein synthesis, and as a consequence, cell growth and proliferation (PubMed:20430826, PubMed:24532714). Stabilizes mRNAs species with a 5'TOP motif, which is required to prevent apoptosis (PubMed:20430826, PubMed:23711370, PubMed:25940091, PubMed:28673543). [UniProt].
Specifications
LARP1 | |
Polyclonal | |
Unconjugated | |
Larp1 | |
0031/08; 0225/29; 1810024J12Rik; 3110040D16Rik; BcDNA:LD02611; CG14065; CG14066; CG42551; CG42551-PA; CG42551-PB; CG42551-PD; CG42551-PE; CG42551-PF; CG42551-PG; CG42551-PH; dlarp; Dmel\CG42551; Dmel_CG42551; drosophila La related protein; KIAA0731; l(3)S003108; l(3)S022529; La related protein; La ribonucleoprotein domain family member 1; La ribonucleoprotein domain family, member 1; La-related protein; la-related protein 1; Larp; Larp1; larp-PA; larp-PB; larp-PD; larp-PE; larp-PF; larp-PG; larp-PH; lethal (3) S003108; lethal (3) S022529; meteor; mKIAA0731; RGD1306683 | |
Rabbit | |
Affinity chromatography | |
RUO | |
23367 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.12 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6PKG0 | |
Larp1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 106-205 of human LARP1 (Q6PKG0). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction