Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LARP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589091
Description
Immunogen sequence: FDYSYGYQEH GERTDQPFQT ELNTSMMYYY DDGTGVQVYP VEEALLKEYI KRQIEYYFSV ENLERDFFLR GKMDEQGFLP ISLIAGFQRV QALTTNLNLI LEALKDSTEV EIVDEKMRKK IEPEKWPIPG PPPRSVPPTD FSQLIDCPEF V; Positive Samples: A-549, HT-29, HeLa, U-87MG, Mouse brain.
LARP2 encodes a protein containing domains found in the La related protein of Drosophila melanogaster. La motif-containing proteins are thought to be RNA-binding proteins, where the La motif and adjacent amino acids fold into an RNA recognition motif. The La motif is also found in proteins unrelated to the La protein. Alternative splicing has been observed at this locus and three variants, encoding distinct isoforms, are described.
Specifications
LARP2 | |
Polyclonal | |
Unconjugated | |
LARP1B | |
1700108L22Rik; 4933421B21Rik; AA690246; DKFZp686L13217; fc76b04; la related protein; La ribonucleoprotein 1B; La ribonucleoprotein domain family member 1B; La ribonucleoprotein domain family member 2; La ribonucleoprotein domain family, member 1B; La ribonucleoprotein domain family, member 2; La-like protein 1B-like protein; la-related protein 1; la-related protein 1B; la-related protein 1B; La-related protein 2; LARP; LARP1B; LARP2; MGC117277; MGC75174; RGD1307509; wu:fc76b04 | |
Rabbit | |
Affinity chromatography | |
RUO | |
214048, 310348, 55132 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.66 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q659C4 | |
LARP1B | |
Recombinant fusion protein containing a sequence corresponding to amino acids 170-320 of human LARP1B (NP_0605482). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction