Learn More
Abnova™ LARP4 293T Cell Transient Overexpression Lysate (Denatured) (T01)
Human overexpression lysate
Manufacturer:  Abnova™ H00113251T01
Catalog No. 89-950-392
Add to cart
Specifications
Specifications
293T
NM_199188
pCMV-LARP4 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LARP4 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 81.2kDa
Transient overexpression cell lysate was tested with anti-LARP4 antibody (Mfr. No. H00113251-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human