Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LARP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109902
Description
Immunogen sequence: MLLFVEQVAS KGTGLNPNAK VWQEIAPGNT DATPVTHGTE SSWHEIAATS GAHPEGNAEL SEDICKEYEV MYSSSCETTR NTTGIEESTD GMILGPEDLS YQIYDVSGES NSAVSTEDLK.
LARP4 gene ontology annotations related to this gene include RNA binding.
Specifications
LARP4 | |
Polyclonal | |
Unconjugated | |
LARP4 | |
c-Mpl binding protein; DXErtd793; DXErtd793e; La ribonucleoprotein domain family member 4; La ribonucleoprotein domain family, member 4; la-related protein 4; la-related protein 4-like; Larp4; PP13296 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100910299, 207214 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.09 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BWW4 | |
LARP4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human LARP4 (NP_443111.4). | |
100 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction