Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LARP4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548876

Catalog No. PA548876

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Plays a role in the regulation of cell morphology and cytoskeletal organization.
Specifications

Specifications

LARP4
Polyclonal
PBS with 0.09% sodium azide
Q71RC2, Q8BWW4
La-related protein 4, La ribonucleoprotein domain family member 4, LARP4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
113251, 207214
Western Blot
Unconjugated
LARP4
Liquid
LARP4
KLH conjugated synthetic peptide between 689-717 amino acids from the C-terminal region of human LARP4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit