Learn More
LARP7 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303724AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The function remains unknown.Specifications
LARP7 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q05CL8, Q4G0J3 | |
Larp7 | |
Between 532 and 582 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
LARP7 | |
ALAZS, DKFZP564K112, HDCMA18P, La ribonucleoprotein domain family member 7, La ribonucleoprotein domain family, member 7, PIP7S, P-TEFb-interaction protein for 7SK stability | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28036, 51574 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.