Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LAS1L Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599097
Description
LAS1L Polyclonal Antibody for Western Blot, ICC/IF, ELISA
Involved in the biogenesis of the 60S ribosomal subunit. Required for maturation of the 28S rRNA. Functions as a component of the Five Friends of Methylated CHTOP (5FMC) complex; the 5FMC complex is recruited to ZNF148 by methylated CHTOP, leading to desumoylation of ZNF148 and subsequent transactivation of ZNF148 target genes.
Specifications
LAS1L | |
Polyclonal | |
Unconjugated | |
Las1l | |
1810030A06Rik; 5830482G23Rik; dJ475B7.2; LAS1 like ribosome biogenesis factor; LAS1 like, ribosome biogenesis factor; LAS1L; Las1-like; LAS1-like (S. cerevisiae); LAS1-like, ribosome biogenesis factor; MSTP060; Protein LAS1 homolog; RGD1565755; Ribosomal biogenesis protein LAS1L | |
Rabbit | |
Protein G | |
RUO | |
81887 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
4.39 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9Y4W2 | |
Las1l | |
Recombinant Human Ribosomal biogenesis protein LAS1L protein (490-565AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction