Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Laser Printable Inserts
For laser printers. Brady Laser Printable Inserts are made of B-124 non adhesive paper and also used as 110 terminal block marking for telecommunications.
Supplier: Brady™ LAT175124RD
Specifications
Red | |
Paper (B-124) | |
Cable and wire marking, Panel labeling, Voice and data identification | |
Rectangle | |
7.77 x 1.27 cm (W x H) |
Matte | |
Laser printable insert | |
Laser Printers | |
3.062 x 0.5 in. (W x H) |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title