Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Lass1 Polyclonal Antibody
Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518992
Description
This antibody is predicted to react with bovine, mouse and porcine based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 64,000.
This gene encodes a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site that is cleaved to produce a mature protein containing seven conserved cysteine residues. Members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in yeast suggest that the encoded protein is involved in aging. This protein is transcribed from a monocistronic mRNA as well as a bicistronic mRNA, which also encodes growth differentiation factor 1.
Specifications
Lass1 | |
Polyclonal | |
Unconjugated | |
CERS1 | |
Ceramide synthase 1; CERS1; EPM8; LAG1; LAG1 homolog, ceramide synthase 1; LAG1 longevity assurance homolog 1; LASS1; longevity assurance (LAG1, S. cerevisiae) homolog 1; longevity assurance gene 1 protein homolog 1; longevity assurance homolog 1; longevity assurance-like protein 1; MGC90349; protein UOG-1; UOG1; upstream of GDF1 | |
Goat | |
Ammonium Sulfate Precipitation | |
RUO | |
10715 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P27544 | |
CERS1 | |
Synthetic peptide sequence (KDLREYDTAEAQ) corresponding to the internal amino acids of LASS1 (aa 318-330). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction