Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LASS2 Mouse anti-Human, Fish, Zebrafish, Clone: B3-2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266930
Description
LASS2 Monoclonal antibody specifically detects LASS2 in Human, Fish, Zebrafish samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).Specifications
LASS2 | |
B3-2 | |
Unconjugated | |
TBS (pH7.4), 0.5%BSA, 40% Glycerol with 0.05% Sodium Azide | |
ceramide synthase 2, CerS2, LAG1 homolog, ceramide synthase 2, LAG1 longevity assurance 2, LAG1 longevity assurance homolog 2, LAG1 longevity assurance homolog 2 (S. cerevisiae), S. cerevisiae) homolog 2, SP260MGC987, TMSG1, Tumor metastasis-suppressor gene 1 protein | |
Mouse | |
IgG2b | |
100μL | |
Primary | |
29956 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
2 mg/mL | |
Western Blot 1:500-1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Purified | |
CERS2 | |
Peptide | |
Protein A purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Fish, Human, Zebrafish |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title