Learn More
LAT Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0793-17R34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP236788AF350
Catalog No. NP236788A30
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot.
Specifications
Specifications
LAT
MM0793-17R34
Western Blot
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human LAT with pY161 in WBs
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
LAT
Phosphopeptides with Y161 of human LAT
0.1 mL
Primary
27040
Human