Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LAT Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0793-17R34, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236788AF594

Catalog No. NP236788A94


Add to cart

Description

Description

LAT Monoclonal antibody specifically detects LAT in Human samples. It is validated for Western Blot.
Specifications

Specifications

LAT
MM0793-17R34
Western Blot
Conjugated, Purified
LAT
Phosphopeptides with Y161 of human LAT
Protein G purified
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary
27040.0
Human
Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human LAT with pY161 in WBs
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit