Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LAT Mouse anti-Human, Mouse, Clone: 11B.12, Novus Biologicals™
Mouse Monoclonal Antibody
$176.84 - $608.00
Specifications
Antigen | LAT |
---|---|
Clone | 11B.12 |
Dilution | Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
LAT Monoclonal specifically detects LAT in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
LAT | |
Western Blot 1:100-1:2000, Immunoprecipitation 1:10-1:500 | |
Monoclonal | |
Mouse | |
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction | |
O43561 | |
27040 | |
GST fusion protein corresponding to residues 31-233 of human LAT, cytoplasmic domain. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
11B.12 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
Human, Mouse | |
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36 | |
LAT | |
IgG1 | |
Affinity Purified | |
LAT 36/38kDa weight may be detected. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title