Learn More
LAT (p Tyr161) Mouse anti-Human, DyLight 755, Clone: MM0793-17R34, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236788IR
Catalog No. NBP236788IR
Add to cart
Description
Description
LAT p Tyr161 Monoclonal antibody specifically detects LAT p Tyr161 in Human samples. It is validated for Western Blot.
Specifications
Specifications
LAT (p Tyr161)
MM0793-17R34
Western Blot
Purified
LAT
Phosphopeptides with Y161 of human LAT
Protein G purified
Adaptive Immunity, Immunology, Phospho Specific, Signal Transduction
Primary
27040
Human
Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
36 kDa phospho-tyrosine adapter protein, LAT1,36 kDa phospho-tyrosine adaptor protein, linker for activation of T cells, linker for activation of T-cells family member 1, p36-38, pp36
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human LAT with pY161 in WBs