Learn More
Enzo Life Sciences Latanoprost (5 mg). CAS: 130209-82-4
SDP
Supplier:  Enzo Life Sciences BMLRA1200005
Encompass_Preferred
Prodrug (isopropyl ester) of 17-phenyl-13,14-dihydro-prostaglandin F2α. The free acid form is a potent agonist at FP receptors. It is in clinical use as a topical antiglaucoma agent. Purity: ≥98% (Assay). Long Term Storage: -20°C.
Catalog No. 50-201-1728
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.