Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latanoprost, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
130209-82-4
C26H40O5
432.601
GGXICVAJURFBLW-WRBHGYNOSA-N
isopropyl 7-1r,2r,3r,5s-3,5-dihydroxy-2-r-3-hydroxy-5-phenylpentyl cyclopentyl hept-5-enoate
86769173
propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]hept-5-enoate
CC(C)OC(=O)CCCC=CCC1C(CC(C1CCC(CCC2=CC=CC=C2)O)O)O
This item is not returnable. View return policy
130209-82-4
C26H40O5
432.601
GGXICVAJURFBLW-WRBHGYNOSA-N
isopropyl 7-1r,2r,3r,5s-3,5-dihydroxy-2-r-3-hydroxy-5-phenylpentyl cyclopentyl hept-5-enoate
86769173
propan-2-yl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-(3-hydroxy-5-phenylpentyl)cyclopentyl]hept-5-enoate
CC(C)OC(=O)CCCC=CCC1C(CC(C1CCC(CCC2=CC=CC=C2)O)O)O

FP prostaglandin receptor agonist; also exhibits agonist activity at EP1 and EP3 receptors

FP prostaglandin receptor agonist; displays greater selectivity than prostanglandin F2α (Cat. No. 4214). Exhibits agonist activity at EP1 and EP3 receptors, but little-to-no effect at EP2, DP, IP and TP prostanoid receptors.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 432.59
Percent Purity >97%
Chemical Name or Material Latanoprost

Recommended Storage : Store at -20°C