Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latent TGF-beta bp1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP30342920UL

 View more versions of this product

Catalog No. NB095470


Add to Cart

Description

Description

Latent TGF-beta bp1 Polyclonal specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Latent TGF-beta bp1
Polyclonal
PBS with 50% glycerol, pH7.3.
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
Unconjugated
latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 1290-1540 of human Latent TGF-beta bp1 (NP_996826.2).
20 μL
Ovarian Carcinoma Cell Markers
4052
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit