Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latent TGF-beta bp1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Latent TGF-beta bp1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095469
View Documents
Novus Biologicals
NBP303429100UL
100 μg Each for $320.71
Add to cart
 
NB095470
View Documents
Novus Biologicals
NBP30342920UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Latent TGF-beta bp1 Polyclonal antibody specifically detects Latent TGF-beta bp1 in Human samples. It is validated for Western Blot
Specifications

Specifications

Latent TGF-beta bp1
Unconjugated
Rabbit
Ovarian Carcinoma Cell Markers
PBS with 50% glycerol, pH7.3.
4052
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human
latent transforming growth factor beta binding protein 1, latent-transforming growth factor beta-binding protein 1, LTBP-1, MGC163161, TGF-beta1-BP-1, Transforming growth factor beta-1-binding protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 1290-1540 of human Latent TGF-beta bp1 (NP_996826.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit