Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ LATEX™ 400nm Blue Midi Antibody Conjugation Kit

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to high quality 400 nm blue latex beads quickly and easily.

Manufacturer:  Novus Biologicals™ 10000120

Catalog No. 10000120


Add to cart

Description

Description

The LATEX™ Conjugation Kit allows antibodies or proteins to be conjugated to our high quality 400nm blue latex beads quickly and easily. The latex beads are designed for ease of use and have much improved handling compared with traditional latex. This has been achieved by specially treating the particles. The conjugation is covalent, and requires less antibody than traditional passive and covalent latex conjugates.

Kit Contents:

  • 1 vial of 10x Quencher buffer
  • 1 vial of Resuspension buffer
  • 1 vial of Reaction buffer A
  • 1 vial of Reaction buffer B
  • 1 Midi vial of 400 nm Blue LATEX
Specifications

Specifications

40 μg
LATEX™ 400nm Blue Midi
Safety and Handling

Safety and Handling

SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit