Learn More
Fisherbrand™ Latex Cohesive Wrap Bandages
Excellent pressure bandages that do not cut off circulation
 View more versions of this product
Catalog No. 22-270-337
Add to cart
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
This item is not returnable. View return policy
Description
Description
  • Cohesive and flexible while providing excellent porosity
  • Consistent compression with nonslip support
  • Maintain original size and do not shrink in width
  • Adhere to themselves — no clips or fasteners needed
  • Quick, easy removal without scissors
  • Length: 5 yd. (4.6m)
Order Info
Multicolor packs include an assortment of neon green, neon pink, purple, red, light blue, dark blue and many more colors.
Specifications
Specifications
Non-woven Spandex
5 yd.
3.8 cm
5 yd. x 1.5 in. (4.5 m x 3.8 cm)
Excellent pressure bandages that do not cut off circulation, cohesive and flexible while providing excellent porosity, consistent compression with nonslip support
No
Wrap Bandage
4.5 m
1.5 in.
Yes
Tan
CE