Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Latexin Mouse anti-Human, Clone: OTI1E10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525742

Catalog No. PIMA525742


Add to cart

Description

Description

Latexin, the only endogenous specific inhibitor known for zinc-dependent metallocarboxypeptidases, is a 222 amino acid protein belonging to the protease inhibitor I47 (latexin) family. Human latexin is an elongated molecule with N- and C-terminal domains that each consist of an alpha helix enveloped by a curved beta sheet and the two domains are separated by a connecting segment. Latexin is a hardly reversible, non-competitive, and potent inhibitor of CPA1, CPA2 and CPA4 but is less potent against other mammalian carboxypeptidases, and it does not inhibit enzymes of other carboxypeptidase classes like metallopeptidases or serine proteases. In rat, Latexin may be used as a molecular marker for the regional specification of the neocortex. It is expressed in most tissues with high expression in brain, lung and digestive tract.
Specifications

Specifications

Latexin
Monoclonal
0.54 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BS40
Latexin; Endogenous carboxypeptidase inhibitor; ECI; Protein MUM; Tissue carboxypeptidase inhibitor; TCI; LXN
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
56925
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI1E10
Unconjugated
LXN
Liquid
LXN
Full length human recombit protein of LXN produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit