Learn More
Promega LATS1-NanoLuc Fusion Vector, 20microg
The NanoLuc Kinase Fusion Vectors contains the coding region of the indicated human kinase fused to either the C- or the N-terminus of NanoLuc luciferase. Designed for use with the NanoBRET Target Engagement (TE) Intracellular Kinase Assay.
Manufacturer:  Promega NV1501
Catalog No. PRNV1501
Add to cart
Description
Description
  • Kinase Fusion Vector for NanoBRET™ Target Engagement
  • Transfection-ready DNA to express full-length LATS1 kinase fused to NanoLuc(R) luciferase
  • Measures kinase target engagement in live cells
  • Use with NanoBRET™ TE Intracellular Kinase Assay, K-5
Specifications
Specifications
-30°C to -10°C
20 μg