Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LATS2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LATS2
Applications Immunofluorescence, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB092839
View Documents
Novus Biologicals
NBP303913100UL
100 μg
Each for $320.71
Add to cart
 
NB092840
View Documents
Novus Biologicals
NBP30391320UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

LATS2 Polyclonal antibody specifically detects LATS2 in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

LATS2
Unconjugated
Rabbit
Apoptosis, Cancer, Cell Cycle and Replication, GPCR, Tumor Suppressors
PBS with 50% glycerol, pH7.3.
26524
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, Western Blot
Purified
RUO
Human, Mouse
EC 2.7.11, EC 2.7.11.1, FLJ13161, Kinase phosphorylated during mitosis protein, KPM, Large tumor suppressor homolog 2, LATS (large tumor suppressor, Drosophila) homolog 2, LATS, large tumor suppressor, homolog 2 (Drosophila), serine/threonine kinase KPM, Serine/threonine-protein kinase kpm, serine/threonine-protein kinase LATS2, Warts-like kinase
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human LATS2 (NP_055387.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit