Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Lauroylcarnitine chloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
14919-37-0
C19H38ClNO4
379.966
PDBBUDRTWRVCFN-UHFFFAOYSA-N
dl-lauroylcarnitine chloride,lauroyl-dl-carnitine chloride,+/--lauroylcarnitine chloride,c19h38no4.cl,lauroylcarnitine chloride,3-carboxy-2-dodecanoyloxy propyl trimethylazanium chloride,lauric acid, ester with 3-carboxy-2-hydroxypropyl trimethylammonium chloride,3-carboxy-2-hydroxypropyl trimethylammonium chloride, laurate,2-dodecanoyloxy-4-hydroxy-4-oxobutyl-trimethylazanium chloride,ammonium, 3-carboxy-2-hydroxypropyl trimethyl-, chloride, laurate ester
177509
(3-carboxy-2-dodecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]
This item is not returnable. View return policy
14919-37-0
C19H38ClNO4
379.966
PDBBUDRTWRVCFN-UHFFFAOYSA-N
dl-lauroylcarnitine chloride,lauroyl-dl-carnitine chloride,+/--lauroylcarnitine chloride,c19h38no4.cl,lauroylcarnitine chloride,3-carboxy-2-dodecanoyloxy propyl trimethylazanium chloride,lauric acid, ester with 3-carboxy-2-hydroxypropyl trimethylammonium chloride,3-carboxy-2-hydroxypropyl trimethylammonium chloride, laurate,2-dodecanoyloxy-4-hydroxy-4-oxobutyl-trimethylazanium chloride,ammonium, 3-carboxy-2-hydroxypropyl trimethyl-, chloride, laurate ester
177509
(3-carboxy-2-dodecanoyloxypropyl)-trimethylazanium;chloride
CCCCCCCCCCCC(=O)OC(CC(=O)O)C[N+](C)(C)C.[Cl-]

Intermediate in lipid metabolism

Homolog of acetylcarnitine chloride (Cat. No. 0355). Acylcarnitines are important intermediates in lipid metabolism.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 379.97
Percent Purity >98%
Chemical Name or Material (±)-Lauroylcarnitine chloride

Recommended Storage : Desiccate at Room Temperature