Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Mouse anti-Human, Clone: MM0448-4F33, Novus Biologicals™
Mouse Monoclonal Antibody
$177.16 - $493.00
Specifications
Antigen | Layilin |
---|---|
Clone | MM0448-4F33 |
Concentration | LYOPH |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA.Specifications
Layilin | |
LYOPH | |
Unconjugated | |
Mouse | |
Human | |
FLJ30977, FLJ31092, layilin | |
LAYN | |
Protein G purified |
MM0448-4F33 | |
Monoclonal | |
Purified | |
RUO | |
NP_849156 | |
143903 | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title