Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Layilin Rabbit anti-Human, Clone: 107, Novus Biologicals™
Rabbit Monoclonal Antibody
$321.00 - $493.00
Specifications
Antigen | Layilin |
---|---|
Clone | 107 |
Dilution | ELISA 1:5000-1:10000 |
Applications | ELISA |
Classification | Monoclonal |
Description
Layilin Monoclonal specifically detects Layilin in Human samples. It is validated for ELISA.Specifications
Layilin | |
ELISA 1:5000-1:10000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
143903 | |
IgG | |
Protein A purified |
107 | |
ELISA | |
Unconjugated | |
Rabbit | |
Human | |
FLJ30977, FLJ31092, layilin | |
Recombinant Human Layilin protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title