Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LB Broth (Miller's modification), Alfa Aesar™

For the growth of E. coli and other enteric bacteria. LB Luria contains less salt than LB Miller and LB Lennox. LB Luria is used for cultivating salt-sensitive strains of E. coli and enteric bacteria.

$15.10 - $56.70

Specifications

Format Powder
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AAH2667622 View Documents Alfa Aesar
H2667622
100g Glass Bottle Each for $15.10
Order in Bulk

Request bulk or custom formats

Add to cart
 
AAH2667636 View Documents Alfa Aesar
H2667636
500g Plastic Bottle Each for $56.70
Order in Bulk

Request bulk or custom formats

Add to cart
 
Specifications

Specifications

Powder
Safety and Handling

Safety and Handling

Recommended Storage : Ambient temperatures

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit