Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LBP Rat anti-Mouse, Alexa Fluor™ 594, Clone: RM0370-11B31, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP232730AF594

Catalog No. NP232730A94


Add to cart

Description

Description

LBP Monoclonal antibody specifically detects LBP in Mouse samples. It is validated for ELISA.
Specifications

Specifications

LBP
RM0370-11B31
ELISA
Conjugated, Purified
LBP
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with recombinant mouse LBP
Protein G purified
Phospho Specific
Primary
3929.0
Mouse
ELISA
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233
Rat
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human LBP by direct ELISA
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit