Learn More
LBP Rat anti-Mouse, Alexa Fluor™ 750, Clone: RM0370-11B31, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232730AF750
Catalog No. NP232730A70
Add to cart
Description
Description
LBP Monoclonal antibody specifically detects LBP in Mouse samples. It is validated for ELISA.
Specifications
Specifications
LBP
RM0370-11B31
ELISA
lipopolysaccharide binding protein, lipopolysaccharide-binding protein, LPS-binding protein, MGC22233
Rat
IgG1
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human LBP by direct ELISA
ELISA
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
LBP
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells from a rat immunized with recombinant mouse LBP
0.1 mL
Primary
3929
Mouse