Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LC3A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$440.00 - $516.35

Specifications

Antigen LC3A
Host Species Rabbit
Isotype IgG
Gene Alias anti-LC3B, atg8f Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B monoclonal, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated protein 1 light chain 3 betaMAP1A/MAP1B light chain 3 B, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Target Species Human
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP254740 View Documents Novus Biologicals
NBP254740
100 ul Each for $516.35
Add to cart
 
NBP254741 View Documents Novus Biologicals
NBP254741
100 ul Each for $516.35
Add to cart
 
NBP254742 View Documents Novus Biologicals
NBP254742
100 ul Each for $440.00
Add to cart
 
Description

Description

LC3B Polyclonal antibody specifically detects LC3B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

LC3A
IgG
Human
RUO
Affinity Purified
Primary
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Rabbit
anti-LC3B, atg8f Autophagy-related protein LC3 B, Autophagy-related ubiquitin-like modifier LC3 B, LC3B, lc3b autophagy marker, LC3B ihc, LC3B immunohistochemistry, LC3B immunoprecipitation, LC3B monoclonal, LC3B western blot, lc3-i, ii, MAP1 light chain 3-like protein 2, MAP1A/1BLC3, MAP1A/MAP1B LC3 B, map1lc3b, microtubule associated protein 3 b, microtubule associated protein 3 beta, microtubule-associated protein 1 light chain 3 betaMAP1A/MAP1B light chain 3 B, microtubule-associated proteins 1A/1B light chain 3, microtubule-associated proteins 1A/1B light chain 3B
Immunohistochemistry (Paraffin)
Unconjugated
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit