Learn More
Tocris Bioscience™ LCB 03-0110 dihydrochloride Available on GSA/VA Contract for Federal Government customers only.
Potent c-SRC kinase inhibitor; also inhibits DDR2, BTK and Syk
$217.50 - $910.50
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
559210 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
5592/10
Each for $217.50
Add to cart
559250 View Documents Tocris Bioscience™
5592/50
Each for $910.50
Add to cart
Description
Description
Potent c-Src kinase inhibitor (IC50 = 1.3 nM). Also potent inhibitor of discoidin domain receptor 2 (DDR2) family tyrosine kinases, BTK and Syk. Suppresses LPS-induced activation of macrophages and TGF-β1- induced activation of fibroblasts in vitro. Also inhibits activation of macrophages and fibroblasts and suppresses scar formation in a wound healing model.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.