Learn More
Abnova™ LCK 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00003932T02
Catalog No. 89-950-458
Add to cart
Description
Description
Specifications
Specifications
293T
BC013200.1
pCMV-LCK full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
LCK 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 61.2kDa
Transient overexpression cell lysate was tested with anti-LCK antibody (Mfr. No. H00003932-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human