Learn More
Aviva Systems Biology Corp LCK Antibody - N-terminal region (ARP54525_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54525_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LCK. Alternate names: YT16, p56lck, pp58lck, LSKApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep
Catalog No. 50-014-61232