Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lck Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0796-24Z21, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236790AF350

Catalog No. NP236790A30


Add to cart

Description

Description

Lck Monoclonal antibody specifically detects Lck in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications

Specifications

Lck
MM0796-24Z21
Western Blot, Flow Cytometry
Conjugated, Purified
LCK
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with phosphopeptides containing human LckpY505.
Protein G purified
Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
3932.0
Human
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16
Mouse
IgG2
0.1 mL
Store at 4°C in the dark.
Monoclonal
This antibody recognizes human LcK-pY505 in WBs.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit