Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Lck Mouse anti-Human, Clone: MM0795-71W27, Novus Biologicals™
Mouse Monoclonal Antibody
Specifications
Antigen | Lck (p Tyr394) |
---|---|
Clone | MM0795-71W27 |
Concentration | LYOPH |
Dilution | Western Blot 1:1000-2000, Immunocytochemistry/Immunofluorescence 1:40-100 |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP236789
|
Novus Biologicals
NBP236789 |
0.1 mg | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
Lck p Tyr394 Monoclonal antibody specifically detects Lck p Tyr394 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Lck (p Tyr394) | |
LYOPH | |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
P06239 | |
3932 | |
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with phosphopeptides containing human LckpY394. | |
Primary | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
MM0795-71W27 | |
Western Blot 1:1000-2000, Immunocytochemistry/Immunofluorescence 1:40-100 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16 | |
LCK | |
IgG2 | |
Protein G purified | |
This antibody recognizes human LcK-pY394 in WBs. |
For Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title