Learn More
Lck (p Tyr505) Mouse anti-Human, DyLight 755, Clone: MM0796-24Z21, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236790IR
Catalog No. NBP236790IR
Add to cart
Description
Description
Lck p Tyr505 Monoclonal antibody specifically detects Lck p Tyr505 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
Specifications
Lck (p Tyr505)
MM0796-24Z21
Western Blot, Flow Cytometry
Purified
LCK
This antibody was produced from a hybridoma (mouse myeloma fused with spleen cells) from a mouse immunized with phosphopeptides containing human LckpY505.
Protein G purified
Adaptive Immunity, Cell Cycle and Replication, Immunology, Phospho Specific, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primary
3932
Human
Western Blot, Flow Cytometry
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
EC 2.7.10, EC 2.7.10.2, Leukocyte C-terminal Src kinase, LSK, Lymphocyte cell-specific protein-tyrosine kinase, lymphocyte-specific protein tyrosine kinase, p56(LSTRA) protein-tyrosine kinase, p56lck, p56-LCK, pp58lck, Protein YT16, Proto-oncogene Lck, proto-oncogene tyrosine-protein kinase LCK, T cell-specific protein-tyrosine kinase, T-lymphocyte specific protein tyrosine kinase p56lck, tyrosine-protein kinase Lck, YT16
Mouse
IgG2
0.1 ml
Store at 4C in the dark.
Monoclonal
This antibody recognizes human LcK-pY505 in WBs.