Learn More
LCK (p56lck) Ab-1 Mouse Monoclonal Antibody, Epredia™
Recommended for Western Blotting, Immunoprecipitation (Denatured verified) and Kinase Assays, Epredia™ LCK (p56lck) Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Supplier: Epredia MS164P1ABX
Description
p56lck is a membrane-associated non-receptor protein tyrosine kinase from the Src-family of kinases. The phosphorylation status and the activity of p56lck is regulated by the CD45 tyrosine protein phosphatase. p56lck plays a critical role in T cell development and activation. p56lck gene is localized to a site in the genome which undergoes frequent chromosomal abnormalities in lymphomas and neuroblastomas.
Host Species: Mouse
Clone: 3A5
Isotype: IgG2b/κ
Species Reactivity: Human, Mouse and Rat. Others not known.
Epitope: aa 1-225
Immunogen: Recombinant protein corresponding to aa 1-225 of murine p56lck protein
Molecular Weight: 56kDa
Positive Control: Jurkat cells
Cellular Localization: Cell membrane
Recommended for:
- Western Blotting
- Immunoprecipitation (Denatured verified)
- Kinase Assay
Note:
USA: RUO; Int'l: RUO
Specifications
LCK (p56superscriptlcksuperscript) Ab-1 | |
Monoclonal | |
1mg/mL | |
Mouse | |
100 μL | |
Cancer and Tumor Biology | |
Human, Mouse, Rat | |
IgG2b κ |
Immunoprecipitation, Western Blot | |
3A5 | |
Unconjugated | |
Recombinant protein corresponding to aa 1-225 of murine p56superscriptlcksuperscript protein | |
RUO | |
Primary | |
Concentrated |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.