Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCMT1 Mouse anti-Human, Mouse, Rat, Clone: OTI2C9, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503125

Catalog No. 50-167-4091


Add to cart

Description

Description

Perlecan is an extracellular matrix proteoglycan. It has a large core protein of 400-450 kDa and is often produced with heparan sulfate side chains. Perlecan is found in basement membranes where it contributes to the permeability characteristics, serves as a substrate for vascular cells and binds growth factors involved in vascular remodelling.
Specifications

Specifications

LCMT1
Monoclonal
0.51 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6P4Z6, Q9UIC8
CGI-68, LCMT, PPMT1
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
30949, 361643, 51451
Immunohistochemistry (Paraffin), Western Blot
OTI2C9
Unconjugated
LCMT1
Liquid
LCMT1
Full length human recombit protein of human LCMT1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit