Learn More
Abnova™ LCMT1 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Supplier:  Abnova™ H00051451T01
Catalog No. 89-950-463
Add to Cart
Description
Description
Specifications
Specifications
293T
LCMT1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 36.85kDa
Transient overexpression cell lysate was tested with anti-LCMT1 antibody (Mfr. No. H00051451-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
NM_016309.2
pCMV-LCMT1 full-length
100 μL
Human
Product Suggestions
Product Suggestions