Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN1 Rabbit anti-Human, Polyclonal, Proteintech
SDP

Rabbit Polyclonal Antibody

Supplier:  Proteintech Group Inc 179001AP150UL

Encompass_Preferred

 View more versions of this product

Catalog No. 50-173-1863


May include imposed supplier surcharges.
Only null left
Add to Cart

Description

Description

The protein encoded by this gene belongs to the lipocalin family. Lipocalins are a group of extracellular proteins that are able to bind lipophiles by enclosure within their structures to minimize solvent contact. This protein may bind hydrophobic ligands and inhibit cysteine proteinases. It may also play a role in taste reception.
Specifications

Specifications

LCN1
Polyclonal
Unconjugated
LCN1
LCN1, Lipocalin 1, lipocalin 1 (tear prealbumin), PMFA, Tear lipocalin, Tear prealbumin, Tlc, TP, VEG protein, VEGP, Von Ebner gland protein
Rabbit
Antigen Affinity Chromatography
RUO
3933
-20°C
Liquid
Western Blot
0.41 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P31025
LCN1
LCN1 Fusion Protein Ag12270
150 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.