Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCN12 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA544898
Description
Peptide sequence: GNQFQGEWFV LGLAGNSFRP EHRALLNAFT ATFELSDDGR FEVWNAMTRG Sequence homology: Human: 100%; Mouse: 79%.
LCN12 may play a role in male fertility. LCN12 may act as a retinoid carrier protein within the epididymis.
Specifications
LCN12 | |
Polyclonal | |
Unconjugated | |
LCN12 | |
9230102M18Rik; Epididymal-specific lipocalin-12; Lcn12; lipocalcin 12; lipocalin 12 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
286256 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6JVE5 | |
LCN12 | |
Synthetic peptide directed towards the N-terminal of human LCN12 (aa 36-85). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction