Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCN2 Goat anti-Mouse, Polyclonal , Abnova™
Goat polyclonal antibody raised against mouse Lcn2.
Supplier: Abnova Corporation PAB29690
Specifications
LCN2 | |
Polyclonal | |
In PBS, pH 7.2 (0.09% sodium azide) | |
Lcn2 | |
Native purified mouse Lcn2. | |
100 μg | |
Primary | |
Mouse | |
IgG |
ELISA | |
Unconjugated | |
24p3, AW212229, NGAL | |
Goat | |
Affinity Purified | |
RUO | |
16819 | |
Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction