Learn More
Aviva Systems Biology Corp LCN6 Antibody - middle region (ARP51185_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51185_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LCN6. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LCN6. Alternate names: LCN5, UNQ643, hLcn5ApplicationWB. Species reactivity: Dog, Human
Catalog No. 50-014-60513