Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LCN6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP30489720UL
Description
LCN6 Polyclonal specifically detects LCN6 in Mouse, Rat samples. It is validated for Western Blot.Specifications
LCN6 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
158062 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
Unconjugated | |
epididymal-specific lipocalin LCN6, epididymal-specific lipocalin-6, hLcn5, LCN5, lipocalin 5, lipocalin 6, lipocalin-5, UNQ643 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 25-120 of human LCN6 (NP_945184.1). | |
20 μL | |
Primary | |
Mouse, Rat | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction