Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN6 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen LCN6
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095168
View Documents
Novus Biologicals
NBP304897100UL
100 μg Each for $320.71
Add to cart
 
NB095169
View Documents
Novus Biologicals
NBP30489720UL
20 μg Each for $138.37
Add to cart
 
Description

Description

LCN6 Polyclonal antibody specifically detects LCN6 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

LCN6
Unconjugated
Rabbit
Mouse, Rat
epididymal-specific lipocalin LCN6, epididymal-specific lipocalin-6, hLcn5, LCN5, lipocalin 5, lipocalin 6, lipocalin-5, UNQ643
Recombinant fusion protein containing a sequence corresponding to amino acids 25-120 of human LCN6 (NP_945184.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
158062
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit