Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LCN8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548792

Catalog No. PA548792


Add to cart

Description

Description

May play a role in male fertility. May act as a retinoid carrier protein within the epididymis.
Specifications

Specifications

LCN8
Polyclonal
PBS with 0.09% sodium azide
Q6JVE9
Epididymal-specific lipocalin-8, LCN8, LCN5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
138307
Western Blot
Unconjugated
LCN8
Liquid
LCN8
KLH conjugated synthetic peptide between 111-140 amino acids from the C-terminal region of human LCN8.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit