Learn More
LCN9 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 237791AP
Catalog No. 50-173-4122
Add to cart
Description
Description
Members of the lipocalin family, such as LCN9, have a common structure consisting of an 8-stranded antiparallel beta-barrel that forms a cup-shaped ligand-binding pocket or calyx. Lipocalins generally bind small hydrophobic ligands and transport them to specific cells (Suzuki et al., 2004 ).
Specifications
Specifications
LCN9
0.29 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WX39
9230102I19Rik, LCN9, lipocalin 9, MUP like lipocalin
Rabbit
IgG
150 μL
-20°C
Primary
392399
Western Blot
Unconjugated
LCN9
Liquid
LCN9
LCN9 Fusion Protein Ag20205
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human