Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ LCOR Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA585015
Description
Immunogen sequence: QQFAAEYTSK NSSTQDPSQP NSTKNQSLPK ASPVTTSPTA ATTQNPVLSK LLMADQDSPL DLTVRKSQSE PSEQD.
LCOR is a transcriptional corepressor widely expressed in fetal and adult tissues that is recruited to agonist-bound nuclear receptors through a single LxxLL motif, also referred to as a nuclear receptor (NR) box.
Specifications
LCOR | |
Polyclonal | |
Unconjugated | |
LCOR | |
3110023F06Rik; A630025C20Rik; C10orf12; KIAA1795; Lcor; ligand dependent nuclear receptor corepressor; ligand-dependent corepressor; Mblk1-related protein 2; Mblk1-related protein-2; mKIAA1795; MLR2; RGD1311704; transcription factor MLR2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
84458 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96JN0 | |
LCOR | |
Recombinant protein corresponding to Human LCOR. Recombinant protein control fragment (Product #RP-108104). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction